Learn More
Synthware™ Chromatography Column with PTFE Stopcock, Fritted Disc
Chromatography Column with PTFE Stopcock and Fritted Disc for flash chromatography.
Supplier:  Synthware™ C363225C
 View more versions of this product
Catalog No. 31500994
Only null left
Explore available promotions
Description
Description
  • General purpose chromatography column with PTFE stopcock.
  • With reinforced beaded top and coarse porosity fritted disc to support column packing.
  • Heavy-wall design increases safety.
Specifications
Specifications
26 mm
2 mm bore
254 mm
Chromatography Column
40 to 80 μm
32 mm
Product Suggestions
Product Suggestions