Learn More
Synthware™ Buchner Funnels with Perforated Glass Plate
With sealed in perforated glass disc to support filter media
Supplier:  Synthware™ F809600
 View more versions of this product
Catalog No. 31501542
Only null left
Explore available promotions
Description
Description
  • Design solves contamination problem with fritted disc and blindness problem with ceramic Buchner funnel
  • Note: Not recommended for filtration where pressure would exceed 15 psi
Specifications
Specifications
600 mL
Clear
Buchner Funnel with Perforated Glass Plate
Borosilicate Glass (3.3)
90mm
Product Suggestions
Product Suggestions