Synthware™ Buchner Funnels with Inner Joint and Perforated Glass Plate
With sealed in perforated glass disc to support filter media
Supplier: Synthware™ F969024
Description
- Lower standard taper inner joint
- Hose connector for connection to vacuum source
- O.D. of hose barb is approx. 10mm; tubing would need to be at least 10mm
Specifications
600 mL | |
Clear | |
24/40 |
Borosilicate Glass 3.3 | |
90mm | |
Buchner Funnels with Inner Joint and Perforated Glass Plate |