Learn More
Synthware™ Allihn Condenser
GREENER_CHOICE
With joints at the top and the bottom
Supplier:  Synthware™ C251200
 View more versions of this product
Catalog No. 31500792
Only null left
Description
Description
  • With standard taper outer joint at top and lower inner drip tip joint
  • Both joints of same size
Specifications
Chemical Identifiers
Borosilicate Glass (3.3)
Allihn Condenser
Distillation
Distillation Head
Specifications
Borosilicate Glass (3.3)
Distillation Head
24/40
Allihn Condenser
Distillation
None
200 mm
Hose connection: 8mm O.D.
Product Suggestions
Product Suggestions