Learn More
Synthware™ Addition Funnels with Pressure Equalizing Arm and PTFE Stopcock
Cylindrical style addition funnel with pressure equalizing arm and PTFE Stopcock
Supplier: Synthware™ F671425
Description
- Top outer and lower inner standard taper joints of same size
Specifications
| 25 mL | |
| Stopcock bore: 2mm | |
| 14/20 | |
| Pressure-equalizing Funnel with PTFE Stopcock |