missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Pressure Equalizing Funnel with Teflon™ Metering Stopcock

Teflon™ adjustable bore metering plug prevents sudden overflow, or non-flow during reagent addition.
Supplier: Synthware™ F611925
Description
- Fully control flow rate with the Teflon™ adjustable bore metering plug.
- Adjustment knob can be turned from full flow to dropwise.
- Prevents sudden overflow or non-flow during reagent addition.
Chemical Identifiers
| Borosilicate Glass (3.3) | |
| Clear | |
| Pressure-equalizing Funnel with Metering Stopcock |
Specifications
| 25 mL | |
| Clear | |
| Liquid Transfer | |
| 0.8 oz. |
| Borosilicate Glass (3.3) | |
| Yes | |
| 19/22 | |
| Pressure-equalizing Funnel with Metering Stopcock |