Learn More
Synthware™ Pressure Equalizing Funnel with Teflon™ Metering Stopcock
Teflon™ adjustable bore metering plug prevents sudden overflow, or non-flow during reagent addition.
Supplier: Synthware™ F614500
Description
- Fully control flow rate with the Teflon™ adjustable bore metering plug.
- Adjustment knob can be turned from full flow to dropwise.
- Prevents sudden overflow or non-flow during reagent addition.
Chemical Identifiers
| Borosilicate Glass (3.3) | |
| Clear | |
| Pressure-equalizing Funnel with Metering Stopcock |
Specifications
| 4 mm | |
| Borosilicate Glass (3.3) | |
| 24/40 | |
| Pressure-equalizing Funnel with Metering Stopcock |
| 500 mL | |
| Clear | |
| 16.9 oz. |