missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthetic Fresh Water, Moderately Hard, Ricca Chemical
207 ppm TDS as Fresh Water Ions, 80 to 100 mg/L Hardness as CaCO3
Supplier: Ricca Chemical Company 86631
Specifications
| Clear colorless | |
| Natural Poly | |
| 962 | |
| Synthetic Fresh Water |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| 4 L | |
| CHEBI:15377 |