Learn More
Synthetic Fresh Water, Hard, Ricca Chemical
415 ppm TDS as Fresh Water Ions, 160 to 180 mg/L Hardness as CaCO3
Supplier:  Ricca Chemical Company 866432
 View more versions of this product
Catalog No. 866432
Only null left
Description
Description
Specifications
Chemical Identifiers
Clear colorless
Liquid
Natural Poly Bottle
Synthetic Fresh Water
Specifications
Clear colorless
Liquid
1 L
1 L
Natural Poly Bottle
Synthetic Fresh Water
Product Suggestions
Product Suggestions