Learn More
Synoptics™ Synbiosis Validation Kit
Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments and include a validation plate, a validation procedure, and a test certificate.
Supplier: Synoptics™ SYNBKITSPIRAL
Description
- Synbiosis Validation Kits are designed for use with ProtoCOL, Protos, and ChromaZona instruments
- Each kit includes a validation plate, a validation procedure, and a test certificate
- Kits available for validation of: Pour Plate Imaging, Count Plate Imaging, Color Count Imaging, Aone Imagine, East Imaging, Spiral Imaging, Opka Imaging, SBA Imaging
Chemical Identifiers
| Validation Kit | |
| None | |
| Plastic, Paper | |
| None |
| None | |
| None | |
| None | |
| Single |
Specifications
| Validation Kit | |
| None | |
| Plastic, Paper | |
| None | |
| Validation of Spiral Imaging | |
| 2 Sector Spiral Validation Plate, Whole Frame Spiral Validation Plate, Spiral Validation Procedure, Spiral Test Certificate |
| None | |
| None | |
| None | |
| Single | |
| ProtoCOL and Protos |