missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ SynaptoRed™ Reagent, Calbiochem™,

Catalog No. 5747995MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5747995MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5747995MG Supplier MilliporeSigma™ Supplier No. 5747995MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 162112-35-8
Molecular Formula C30H45Br2N3
Molecular Weight (g/mol) 607.519
InChI Key AFVSZGYRRUMOFH-UHFFFAOYSA-L
Synonym FM4-64, Frie Mao 4-64, N-(3-Triethylammoniumpropyl)-4-(p-diethylaminophenylhexatrienyl)-pyridinium, 2Br
PubChem CID 90489009
IUPAC Name 3-[4-[(1Z,3E,5Z)-6-[4-(diethylamino)phenyl]hexa-1,3,5-trienyl]pyridin-1-ium-1-yl]propyl-triethylazanium;dibromide
SMILES CCN(CC)C1=CC=C(C=C1)C=CC=CC=CC2=CC=[N+](C=C2)CCC[N+](CC)(CC)CC.[Br-].[Br-]

Specifications

Packaging Glass Bottle
Color Purple
Quantity 5 mg
Physical Form Solid