missing translation for 'onlineSavingsMsg'
Learn More

syn-2-Pyridinealdoxime, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 1193-96-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006354
InChI Key VRGKDCRTYBCJTA-UHFFFAOYSA-N
Synonym pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
PubChem CID 6740772
IUPAC Name 2-(nitrosomethylidene)-1H-pyridine
SMILES O=NC=C1NC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC131790250
SDS View Documents
Thermo Scientific Chemicals
131790250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC131791000
SDS View Documents
Thermo Scientific Chemicals
131791000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1193-96-0
122.13
VRGKDCRTYBCJTA-UHFFFAOYSA-N
6740772
O=NC=C1NC=CC=C1
C6H6N2O
MFCD00006354
pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
2-(nitrosomethylidene)-1H-pyridine

Specifications

1193-96-0
99+%
C6H6N2O
25g
pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
VRGKDCRTYBCJTA-UHFFFAOYSA-N
2-(nitrosomethylidene)-1H-pyridine
6740772
99+%
Authentic
Glass bottle
MFCD00006354
21, I, 288
Solubility in water: 15g/L (20°C). Other solubilities: may decompose,265g/L ethanol (20°C),31% in methanol- 18% in acetone (20°C),0.5% in toluene- 0.5% in gasoline (20°C)
O=NC=C1NC=CC=C1
122.13
122.13
syn-2-Pyridinealdoxime
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only