missing translation for 'onlineSavingsMsg'
Learn More

syn-2-Pyridinealdoxime, 99+%

Catalog No. AC131790250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC131790250 Supplier Thermo Scientific Chemicals Supplier No. 131790250
Only null left

CAS: 1193-96-0 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1193-96-0
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00167166
InChI Key MTFJSAGADRTKCI-UHFFFAOYSA-N
Synonym pyridine-2-aldoxime, picolinaldehyde oxime, 2-pyridinecarbaldehyde oxime, z-pyridine-2-carbaldehyde oxime, 2-nitrosomethylidene-1h-pyridine, 2-pyridinealdoxime, e-picolinaldehyde oxime, 2-hydroxyiminomethyl-pyridine, 2-pyridine aldoxime, 2-pyridinecarboxaldehyde, oxime, e
PubChem CID 6740772
IUPAC Name 2-(nitrosomethylidene)-1H-pyridine
SMILES ON=CC1=CC=CC=N1

Specifications

Melting Point 110°C to 113°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, I, 288
Solubility Information Solubility in water: 15g/L (20°C). Other solubilities: may decompose,265g/L ethanol (20°C),31% in methanol- 18% in acetone (20°C),0.5% in toluene- 0.5% in gasoline (20°C)
Formula Weight 122.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material syn-2-Pyridinealdoxime
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only