Learn More
Electron Microscopy Sciences Sylgard™ 184
Numéro de catalogue. 50366794
Encompass
Same as the Kwik Gard but in its original container. A two-part silicone elastomer, ideal for potting and encapsulating applications. Very low dielectric constant sealing compound used in patch clamping and many other lab applications. After cure, will withstand -55 degree to 200 degree C. Curing time for this silicone 24hrs@25 degree C, and 15 min.@150 degree C.
Numéro de catalogue. 50366794 Fournisseur Electron Microscopy Sciences Code fournisseur 2423610
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Same as the Kwik Gard but in its original container. A two-part silicone elastomer, ideal for potting and encapsulating applications. Very low dielectric constant sealing compound used in patch clamping and many other lab applications. After cure, will withstand -55 degree to 200 degree C. Curing time for this silicone 24hrs@25 degree C, and 15 min.@150 degree C.