missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Syk Inhibitor

Chemical Identifiers

CAS 622387-85-3
Molecular Formula C18H15N3O3S
Synonym Spleen Tyrosine Kinase Inhibitor
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64281LB0
View Documents Promotion Details
Thermo Scientific Chemicals
J64281LB0
5 mg N/A
Only null left
 
Description

Description

A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

622387-85-3
C18H15N3O3S
Spleen Tyrosine Kinase Inhibitor

Specifications

Dark orange
A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor
C18H15N3O3S
Spleen Tyrosine Kinase Inhibitor
Soluble in DMSO
MLKHXLFEYOOYEY-UHFFFAOYSA-N
3-[(1-methyl-1H-indol-3-yl)methylidene]-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
2°C to 8°C
622387-85-3
353.40
Light Sensitive
Solid
CN1C=C(C=C2C(=O)NC3=C2C=C(C=C3)S(N)(=O)=O)C2=CC=CC=C12
353.40
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only