missing translation for 'onlineSavingsMsg'
Learn More

Sphergo Swivel™ Ergonomic Cleanroom Cleaning Kit

Catalog No. 18366485
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
18366485 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18366485 Supplier Sphergo Supplier No. CLN371

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleanroom cleaning kit with individually packaged components, including microfiber pads and mop that's ergonomically designed to reduce strain.

  • For use with Sphergo Swivel Cleaning Tools
  • 47.05 x 4.72 x 3.15 in.
  • Ergonomic mop reduces strain
  • Designed for use in cleanrooms
  • Individually packaged components
  • Contains: ergonomic pole, mop head, 24 microfiber pads

Specifications

Product Type Mopping System
Bristle Material Microfiber
Bristle Type Pad
Color-coded No
Handle Material Aluminum
Handle Type Extension Pole
Scent Unscented
Sterility Non-sterile
Dimensions (L x W x H) 47.05 x 4.72 x 3.15 in.
Disposable Single-use