missing translation for 'onlineSavingsMsg'
Learn More

Sphergo Swivel™ Disposable Cleanroom Cleaning Pad

Catalog No. 18366488
Change view
Click to view available options
Disposable:
Single-use
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable
18366488 Single-use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18366488 Supplier Sphergo Supplier No. CLN382

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable cleaning pad that comes in bag-in-bag packaging.

  • For use with Sphergo Swivel Cleaning Tools
  • 19 x 5 x 0.5 in.
  • Ideal for use in cleanrooms
  • Bag-in-bag packaging for cleanroom application
  • Single-use application to reduce risk of cross-contamination
  • Compatible with chlorine bleach, hydrogen peroxide, and other common disinfectants
  • USP 797 Compliant
  • Bonded edges improve performance greatly, reducing linting for a cleaner clean
  • 300-360 sq ft cleaning coverage
  • Absorbs 9 x its weight in liquid
  • Channels efficiently pick up and retain dirt in pad
  • Embossed pattern reduces friction for less strain on workers
  • Hook-and-loop backing provides a more aggressive hold, making it the ideal choice for walls
  • Eco-friendly manufacturing with no chemical dyes used in the process
  • 50% post-consumer recycled content
  • Quantity: 240 pads

Specifications

Product Type Mopping System
Bristle Material Microfiber
Bristle Type Pad
Color-coded No
Scent Unscented
Sterility Non-sterile
Dimensions (L x W x H) 19 x 5 x 0.5 in.
Disposable Single-use