Learn More
Heidolph™ Switch Reed Contact for Laborota 20,Rotary Evaporator
Catalog No. 13900946
Change view
Click to view available options
For Use With (Equipment):
Laborota 20 Rotary Evaporator Enclosure
Catalog No. For Use With (Equipment)
13900946 Laborota 20 Rotary Evaporator Enclosure
1 options
Catalog No. 13900946 Supplier Heidolph™ Supplier No. 036280060
Switch reed contact monitors whether the hood is open or not.
  • Included with Laborota 20 Rotary Evaporator
  • Includes switch reed contact and magnet switch
Specifications
Type Enclosure
For Use With (Equipment) Laborota 20 Rotary Evaporator Enclosure