missing translation for 'onlineSavingsMsg'
Learn More

Velp™ Switch for ARE 6 Stirrer

Catalog No. 11392002
Change view
Click to view available options
Type:
Power Supply
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
11392002 Power Supply
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11392002 Supplier Velp™ Supplier No. 10007163

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Switch for use with the ARE 6 Stirrer.

  • On/off switch (O/I) for stirrer

Specifications

Color Black
Dimensions (L x W) 19.5 x 13 mm
For Use With (Equipment) ARE 6 Stirrer
Type Power Supply