missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAI Sweat-less Cleanroom Headbands
Sweat-less headbands are for use under a bouffant hat in a clean environment and reduce contamination risk. They are comfortable, soft, absorbent, and disposable.
Supplier: VAI SL01L
Description
- Extremely durable and resistant to liquids
- Latex and lint free
- Non-sterile and available in small, medium, and large sizes
Specifications
| Cleanroom Headband | |
| Nylon | |
| Single-use | |
| SL-01-L | |
| 20 Pk., 10 Pk./Cs. | |
| 40.6 cm |
| Large | |
| White | |
| Yes | |
| Non-sterile | |
| 16 in. | |
| Liquid |