Learn More
Thermo Scientific™ Spring Loaded Transfer Line Nut for ISQ and TSQ
Catalog No. 290BT240
Change view
Click to view available options
For Use With (Equipment):
Swap-IT
Thermo Scientific ISQ and TSQ
Description:
Chromatography Accessories
Spring Loaded Transfer Nut
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Description
290BT240 Swap-IT Chromatography Accessories
03251526 Thermo Scientific ISQ and TSQ Spring Loaded Transfer Nut
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 290BT240 Supplier Thermo Scientific™ Supplier No. 290BT240
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Specifications
For Use With (Equipment) Swap-IT
Description Chromatography Accessories