Learn More
MilliporeSigma™ Supelco™ Swagelok™ Zero-Dead Volume Union Swagelok
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
Supplier:  MilliporeSigma™ Supelco™ 22052
 Afficher plus de versions de ce produit
Numéro de catalogue. 111004734
Il en reste null
Description
Description
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Swagelok™ zero-dead- volume unions are principally designed for liquid and gas chromatography. On application of Swagelok™ zero-dead- volume unions, a perfect low-dead-volume connection between column terminator and injector or detector is obtained.
Spécifications
Identifiants chimiques
1/Pk.
Fitting: 1/16 in.; zero dead volume
Spécifications
Stainless Steel
1/Pk.
Fitting: 1/16 in.; zero dead volume
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Swagelok is a registered trademark of Swagelok Company