Learn More
MilliporeSigma™ Supelco™ Swagelok™ Tee Wrench 1/4 in.
Catalog No. 111004292
Change view
Click to view available options
Swagelok™ Tee Wrench 1/4 in.
Catalog No.
111004292 Swagelok™ Tee Wrench 1/4 in.
1 options
Catalog No. 111004292 Supplier MilliporeSigma™ Supelco™ Supplier No. 21983U
Made specifically for 1/4 in.. Swagelok™ union tees and crosses.
Specifications
For Use With (Application) Chromatography Applications
Swagelok is a registered trademark of Swagelok Company