Learn More
MilliporeSigma™ Supelco™ Swagelok™ Nut, Front and Back Ferrule Set, 200-S, Brass, 1/8 in.
Catalog No. 111020943
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
111020943 10/Pk.
1 options
Catalog No. 111020943 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA22014
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.
Specifications
Packaging Quantity 10/Pk.
Material Brass
Swagelok is a registered trademark of Swagelok Company