Learn More
Terumo™ Surshield™ Safety Winged Infusion Sets
Catalog No. 22272338
Change view
Click to view available options
Needle Gauge:
19 G
21 G
23 G
25 G
Length (English) Tube:
3.5 in.
12 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Tube
22272338 19 G 3.5 in.
22272340 23 G 3.5 in.
22272341 25 G 3.5 in.
22272334 19 G 12 in.
22272335 21 G 12 in.
22272336 23 G 12 in.
22272337 25 G 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22272338 Supplier Terumo™ Supplier No. SVS19BLS
One-handed activation for IV infusion therapy
  • Nonlatex plastic and tubing
  • Allergy-free usage
  • Flexible wings
  • Polyvinyl tubing
  • Siliconized needle
Specifications
Product Type Safety Winged Infusion Set
Material PVC Tubing, Siliconized Needle
Needle Gauge 19 G
Sterility Sterile
For Use With (Application) IV infusion therapy
Length (English) Needle 0.75 in.
Length (English) Tube 3.5 in.