missing translation for 'onlineSavingsMsg'
Learn More

HR Pharmaceuticals Surgilube™ Surgical Lubricant

Catalog No. p-7250482
Encompass
Change view
Click to view available options
Packaging:
Laminated Tube with Flip-Top Cap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
22355176 Laminated Tube with Flip-Top Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22355176 Supplier HR Pharmaceuticals Supplier No. FOU 0281020537

This sterile, surgical lubricant is non-irritating to the skin, tissue, and mucous membranes. It is especially formulated for surgical and gynecological lubrication.

  • Surgilube™ sterile, surgical lubricant may be used where a sterile, water soluble, non-staining lubricating jelly is indicated.
  • Will not affect surgical instruments, rubber or plastics.

Specifications

Product Type Lubricant
Format Paste
For Use With (Equipment) Catheters, endoscopes, speculums, or other surgical instruments for body orifices
Packaging Laminated Tube with Flip-Top Cap
Weight (English) 4.25 oz.