missing translation for 'onlineSavingsMsg'
Learn More

HR Pharmaceuticals Surgilube™ Surgical Lubricant, FOILPAC™

Catalog No. 22355177
Encompass
Change view
Click to view available options
Format:
Paste
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22355177 Paste
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22355177 Supplier HR Pharmaceuticals Supplier No. FOU 0281020543

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This sterile, surgical lubricant is non-irritating to the skin, tissue, and mucous membranes. It is especially formulated for surgical and gynecological lubrication.

  • Surgilube™ sterile, surgical lubricant may be used where a sterile, water soluble, non-staining lubricating jelly is indicated.
  • Will not affect surgical instruments, rubber or plastics.

Specifications

Product Type Lubricant
Format Paste
For Use With (Equipment) Catheters, endoscopes, speculums, or other surgical instruments for body orifices
Description SURGILUBE™ 3gm FOILPAC™ (Foil Laminated Film)