Learn More
Poltex Surgical Shield Mask Holder
Catalog No. 22000472
Change view
Click to view available options
Mounting:
Wall Mount
Catalog No. Mounting
22000472 Wall Mount
1 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22000472 Supplier Poltex Supplier No. SURGMSKW
Only null left
Consumables shown for demonstration purposes only and are not included with the product.
Holds and dispenses one box of surgical shield masks.
  • Holds Surgical Shield mask boxes up and out of the way.
  • Constructed from durable 1/4 in. thick acrylic
  • Comes in two mounting methods: wall mount, or with powerful magnets.
  • Inside Dimensions: 9 in. wide x 7.75 in. tall x 4 in. deep (front to back)
  • Fits Hallyard 47137, 48247 and related
Specifications
Product Type Face Mask Dispenser
Material PETG
For Use With (Application) Healthcare
For Use With (Equipment) Surgical shield masks
Mounting Wall Mount