missing translation for 'onlineSavingsMsg'
Learn More

Surgical Design Surgical Knife Scalpel Handle, Extra Long

Catalog No. 22079770
Change view
Click to view available options
Size:
# 3
# 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22079770 # 4
22079769 # 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22079770 Supplier Surgical Design Supplier No. 271L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Surgical Design™ Surgical Knife Scalpel Handle (Extra Long) used for sharp and precise dissection. The knife handle comes with patterned surface for enhanced comfort and secure grip that increases precision during use.

  • Surgical Design™ Premium products exceed performance and quality expectations. Guaranteed.
  • Handles designed for precision work
  • Comfortable and reusable
  • Blades slide on smoothly and securely
  • Superb quality and workmanship
  • Premium surgical grade stainless steel
  • Corrosion protected by passivation and ultrasonic cleaning
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable

Specifications

Blade Material Stainless Steel
Product Type Scalpel Handle
Size # 4
Length (English) 8.5 in.
Length (Metric) 21.59 cm
For Use With (Equipment) Compatible with 20 Series Disposable Scalpel Blade (Sizes # 20 to # 25)
Description Satin, Anti-glare
Disposable Reusable
Autoclavable Autoclavable
Color Silver