Learn More
World Precision Surgical Instrument Lubricant DFS Item
Supplier:  World Precision WP500126
  • 8oz (240ml)
  • Anticorrosive
  • Steam Compatable
  • non-greasy
  • prevent spotting,rusting or staining
  • Non-Toxic
  • The lubricant should be applied after cleaning and before sterilizing instruments. This is a non-silicone based surgical instrument lubricant, which is steam permeable, non-greasy and anti-corrosive.
Catalog No. 50822368
Only null left
Explore available promotions