missing translation for 'onlineSavingsMsg'
Learn More

FEATHER Surgical Handle DF-160S-3

Catalog No. 361031009
Change view
Click to view available options
Length (Metric):
160 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
361031009 160 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361031009 Supplier FEATHER Supplier No. DF1031009

FEATHER Surgical Handles are available to meet diverse needs. Each is scientifically designed for smooth loading, functional handling ease and perfect balance. The handles assure long life service and the blade integrates into the handle to form a single instrument.

  • Made in Japan
  • With smooth round shape, weight balance, and ideal length, this handle provides operability for incision in fine or deep areas

Specifications

Blade Material Titanium Alloy with DLC Coating
Product Type Surgical Handle
Size 160 mm
Length (Metric) 160 mm
For Use With (Equipment) FEATHER Surgical Blade # 10, # 11, # 12, # 12d, # 14, # 15, # 15s, # 15c, # 350, # 370, # 390, # 391, # 390C
Disposable Reusable
Sterility Non-sterile
Autoclavable Autoclavable
Quantity 1/Pk.