Learn More
FEATHER Surgical Blade (Microsurgery)
Catalog No. 361031011
Change view
Click to view available options
Size:
# 370
# 390
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
361031011 # 390
361031010 # 370
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361031011 Supplier FEATHER Supplier No. NO. 390
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FEATHER surgical blades No.350, 370, 390, 390C and 391 are small shaped surgical blades, suitable for microsurgery (fine incision) with microscope. With appropriate shape and thickness, these blades are easy to control and use for fine incisions.
  • Made in Japan
  • Individually packed
  • Single use only
Specifications
Blade Material Stainless Steel
Size # 390
For Use With (Equipment) FEATHER Surgical Handle # 3, # 3L, # 7, or # DF-160S-3
Disposable Single-use
Sterility Sterile
Certifications/Compliance ISO13485:2016
Quantity 10/Pk.