Learn More
FEATHER Surgical Blade
Catalog No. 089185E
Change view
Click to view available options
Size:
# 24
# 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
089185E # 24
089185F # 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 089185E Supplier FEATHER Supplier No. NO.24
FEATHER Surgical Blade Stainless Steel is available in a variety of sizes to satisfy a wide range of surgical demands.
  • Made in Japan
  • Individually packed
  • Single use only
Specifications
Blade Material Stainless Steel
Product Type Surgical Blade
Size # 24
For Use With (Equipment) FEATHER Surgical Blade Handles # 4 and # 4L
Disposable Single-use
Sterility Sterile
Certifications/Compliance ISO13485:2016
Quantity 10/Pk.