Learn More
PURELL™ Surface Sanitizer and Disinfectant Battery Powered Sprayer
Catalog No. 19270254
Change view
Click to view available options
Product Type:
Battery-powered Sprayer
Catalog No. Product Type
19270254 Battery-powered Sprayer
1 options
Catalog No. 19270254 Supplier PURELL™ Supplier No. 535004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Battery powered sprayer for PURELL™ surface sanitizer and disinfectant sprays.
  • Handheld battery powered sprayer for easy and efficient disinfection of larger surfaces
  • Simple, one-button operation makes big disinfecting jobs easy
  • Adjustable spray-pattern nozzle
  • Fold-up spray wand and holster for easy storage
  • Battery Operated with 4 x AA Batteries
  • Quickly attaches to 1 gallon bottles of PURELL™ Surface Sanitizing and Disinfectant Spray
  • Ready to Use - No mixing or measuring required
Specifications
Product Type Battery-powered Sprayer
For Use With (Equipment) PURELL™ Surface Sanitizing and Disinfectant Spray