Learn More
Thermo Scientific™ SURF-CAL™ Particle Size Standards
Simplify the preparation of calibration wafers with these particle size standards.
$816.19 - $5,819.25
Specifications
Additive None
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Contents Polystyrene microspheres in deionized, filtered water
For Use With (Application) SSIS Calibration and Maintenance
Material Polystyrene
View More Specs
Products 33
Catalog Number Mfr. No. Diameter (Metric) Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Concentration Price Quantity & Availability  
09980138
View Documents Promotion Details
Thermo Scientific™
PD047
0.047 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980139
View Documents Promotion Details
Thermo Scientific™
PD047B
0.047 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980140
View Documents Promotion Details
Thermo Scientific™
PD064
0.064 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980141
View Documents Promotion Details
Thermo Scientific™
PD064B
0.064 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980146
View Documents Promotion Details
Thermo Scientific™
PD083
0.083 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980147
View Documents Promotion Details
Thermo Scientific™
PD083B
0.083 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980150
View Documents Promotion Details
Thermo Scientific™
PD092
0.092 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980151
View Documents Promotion Details
Thermo Scientific™
PD092B
0.092 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980152
View Documents Promotion Details
Thermo Scientific™
PD100
0.100 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980153
View Documents Promotion Details
Thermo Scientific™
PD100B
0.100 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980178
View Documents Promotion Details
Thermo Scientific™
PD1100
1.112 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980179
View Documents Promotion Details
Thermo Scientific™
PD1100B
1.112 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980157
View Documents Promotion Details
Thermo Scientific™
PD125B
0.126 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980158
View Documents Promotion Details
Thermo Scientific™
PD155
0.155 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980159
View Documents Promotion Details
Thermo Scientific™
PD155B
0.155 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980162
View Documents Promotion Details
Thermo Scientific™
PD200
0.202 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980181
View Documents Promotion Details
Thermo Scientific™
PD2000
2.01 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980163
View Documents Promotion Details
Thermo Scientific™
PD200B
0.202 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980164
View Documents Promotion Details
Thermo Scientific™
PD204
0.204 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980165
View Documents Promotion Details
Thermo Scientific™
PD204B
0.204 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980166
View Documents Promotion Details
Thermo Scientific™
PD215
0.220 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980167
View Documents Promotion Details
Thermo Scientific™
PD215B
0.220 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980183
View Documents Promotion Details
Thermo Scientific™
PD3000
3.04 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980168
View Documents Promotion Details
Thermo Scientific™
PD305
0.304 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980169
View Documents Promotion Details
Thermo Scientific™
PD305B
0.304 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980170
View Documents Promotion Details
Thermo Scientific™
PD365
0.360 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980171
View Documents Promotion Details
Thermo Scientific™
PD365B
0.360 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980172
View Documents Promotion Details
Thermo Scientific™
PD500
0.498 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980173
View Documents Promotion Details
Thermo Scientific™
PD500B
0.498 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
09980174
View Documents Promotion Details
Thermo Scientific™
PD800
0.809 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980175
View Documents Promotion Details
Thermo Scientific™
PD800B
0.809 μm 1 x 10¹° particles/mL
Each for $5,819.25
Only null left
09980176
View Documents Promotion Details
Thermo Scientific™
PD802
0.802 μm 3 x 108 particles/mL
Each for $816.19
Only null left
09980177
View Documents Promotion Details
Thermo Scientific™
PD802B
0.802 μm 1 x 10¹° particles/mL
Each for $5,479.50
Only null left
Description
Description
Simplify the preparation of calibration wafers with Thermo Scientific™ SURF-CAL™ Particle Size Standards, for calibration and maintenance of Scanning Surface Inspection Systems (SSIS). Available particle sizes correspond to the calibration point sizes required by instrument manufacturers.
Thermo Fisher Scientific™, along with the Semiconductor Industry and SEMI, have established specific particle sizes to be used when calibrating Scanning Surface Inspection Systems. Working with instrument manufacturers, we created the SURF-CAL™ product line to meet SEMI standard guidelines.
  • Available sizes are considered to be critical sizing nodes as defined by the International Technology Roadmap for Semiconductors (ITRS).
  • By depositing SURF-CAL™ NIST™ traceable polystyrene latex spheres (PSL) on specially selected wafers, you can perform periodic calibration checks and compare your scanner with scanners at other locations.
  • You can also assess the performance of your SSIS at critical stages in the manufacturing process.
  • All products are suspended in deionized, filtered water in 50mL bottles at a concentration of 3 × 108 particles per mL.
Note:
Products PD1100 and smaller are also available at 1010 particles per mL for applications using the aid of a Differential Mobility Analyzer (DMA) or other size exclusionary techniques.
Specifications
Specifications
None
Polystyrene microspheres in deionized, filtered water
Polystyrene
Certificate of Calibration and Traceability to NIST
50 mL
Product stable for a minimum of 12 months
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
SSIS Calibration and Maintenance
Size Standard Particles
1.05g/cm³
1.59 at 589nm (25°C)