missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Micropoint Pipette Tips, Universal Extended Length
GREENER_CHOICE

Catalog No. 02707455
Change view
Click to view available options
Format:
Bulk
Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
02707455 Bulk
02707469 Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02707455
Get an Extra Pack of Pipette Tips with Purchase
View all promotions

New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips

All nonsterile bulk nonfiltered tips are certified free of RNase, DNase, and DNA.

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Length (English) 1.25 in.
Length (Metric) 46 mm
Clean Claims DNase, RNase, and DNA Free
Format Bulk
Tip Style Extended Length
Packaging Bag of 1000 tips
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Material Polypropylene
Graduations 2.5μL
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.