Learn More
Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
 View more versions of this product
Catalog No. 02-707-471
Only null left
Description
Description
Choose from a comprehensive line of pipet tips for demanding liquid-handling applications
Universal fit for research grade pipetters.
  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable
Specifications
Specifications
30 to 300 μL
Sterile
Not Autoclavable
58 mm
Micro Point
Yes
10 μL, 50 μL, 100 μL, 200 μL
Filtered
Clear
2 in.
Hinged Rack
Universal Fit
SureOne
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.