missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Aerosol Barrier Pipette Tips
GREENER_CHOICE

Catalog No. 02707439
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Tip Style:
Beveled
Extended Length, Micro Point, Low Retention
Micro Point
Micro Point, Extended Length
Color:
Clear
Natural
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style Color
02707439 0.1 to 10 μL Micro Point Natural
02707442 0.1 to 10 μL Micro Point, Extended Length Natural
02707470 0.1 to 20 μL Extended Length, Micro Point, Low Retention Clear
02707432 2 to 20 μL Beveled Natural
02707431 10 to 100 μL Beveled Natural
02707430 20 to 200 μL Micro Point Natural
02707471 30 to 300 μL Micro Point Clear
02707404 100 to 1000 μL Micro Point Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 02707439
Get Three Cases of Pipette Tips with Kit Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose from a comprehensive line of pipet tips for demanding liquid-handling applications

Universal fit for research grade pipetters.

  • Engineered for increased accuracy and are compatible with single-channel, multichannel, and electronic pipetters
  • DNase-/RNase-free; sterile tips are also pyrogen-free
  • Sterile and clear color
  • Filter eliminates aerosol cross-contamination without sealing or trapping samples
  • Tips allow for noncontaminated sample recovery
  • Not autoclavable

Specifications

Volume (Metric) 0.1 to 10 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Length (English) 1.25 in.
Length (Metric) 31 mm
Format Hinged Rack
Tip Style Micro Point
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line SureOne
Graduations 2 μL
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.