missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Stacking Buffer DFS Item

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

Supplier:  Invitrogen™ HC2115

 View more versions of this product

Catalog No. HC2115


Only null left
Explore available promotions

Description

Description

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Specifications

Specifications

5 x 500 mL (2.5 L total) SureCast Stacking Buffer packs.

Store at room temperature.
Room Temperature
Stacking Buffer
5 x 500 mL
SureCast
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.