missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SureCast™ Stacking Buffer
Description
- Pour polyacrylamide gels easily and confidently
- For use with the SureCast vertical gel system
- SureCast gels are designed for use in the Mini Gel Tank
Recommended Storage
Room Temperature
Specifications
Specifications
| Content And Storage | 2 x 500 mL (1 L total) SureCast Stacking Buffer packs. Store at room temperature. |
| Quantity | 2 x 500 mL |
| Shipping Condition | Room Temperature |
| Product Line | SureCast |
| Product Type | Stacking Buffer |
Frequently Asked Questions (FAQs)
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
For Research Use Only. Not for use in diagnostic procedures.