Learn More
Invitrogen™ SureCast™ Gel Handcast Bundle B - Hardware Only
Catalog No. HC1000S
Encompass
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
HC1000S 1 kit
1 options
Catalog No. HC1000S Supplier Invitrogen™ Supplier No. HC1000S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
The SureCast Handcast Bundle B includes all the hardware you need to get started:
  • 2 SureCast Handcast Stations
  • 2 SureCast™ Sealing Pads
  • 2 SureCast™ Glass Plate Sets (2 front and 2 back)
  • 6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
  • 10 SureCast™ Gel Spacers
Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle B. The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.
  • Innovative, leak-free design—minimizes the number of failed gels, resulting in less wasted time
  • Superior glass plate durability
  • Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
  • Unique tilt feature for easy loading of acrylamide solutions
The complete product includes the SureCast™ Handcast System. For a bundle that also includes the SureCast™ Handcast Reagents, please see SureCast™ Gel Handcast Bundle A. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during hand casting. SureCast gels are designed for use in the Mini Gel Tank.
Specifications
Quantity 1 kit
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
For Research Use Only. Not for use in diagnostic procedures.