missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ SUPRO MAX™ Hand Cleaner Refill

Catalog No. 19158121
Change view
Click to view available options
Volume (Metric):
5000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19158121 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19158121 Supplier GOJO™ Supplier No. 7572 02

Fast and effective heavy-duty hand cleaner with gentle scrubbers. Refill is gentle on skin and leaves them feeling healthy.

  • Fast and effective heavy-duty hand cleaner with gentle scrubbers
  • Unique blend of cleaners and skin conditioners help leave skin feeling healthy
  • Non-drying formula
  • SANITARY SEALED™ refill helps prevent contamination
  • Fresh dispensing valve with each refill
  • Effective on oil, grease, paint, and adhesives

Specifications

Product Type Hand Cleanser
Color Beige
Form Lotion
For Use With (Application) Effective on oil, grease, paint, and adhesives.
For Use With (Equipment) 7500-01
Quantity 2/Pk.
Size 5000 mL
Volume (Metric) 5000 mL