Learn More
3M™ Supplied Air Hose, W-9435-100
Catalog No. 19021634
Change view
Click to view available options
1/Pk.
Catalog No.
19021634 1/Pk.
1 options
Catalog No. 19021634 Supplier 3M™ Supplier No. 7000005375
Only null left
Is 100 feet long, equipped with Industrial Interchange MPT fittings at both ends and accepts 1/4 in (0.63 cm) FPT fittings.
  • Supplied air respirators require a source of clean breathing air such as plant air systems, low pressure pumps, or bottled air
  • Complete systems include a supplied air hose, an air regulating valve, a breathing tube, and a helmet, hood, or other facepiece
  • Delve into 3M™ Products and experience the wide variety of products and replacement parts offered with powered air purifying respirators (PAPR)
  • In addition to powered systems, 3M also offers supplied air respirators (SAR) using air from an air tank or a different external source
Specifications
Model W9435100
Quantity 1/Pk.
Diameter (English) 3/8 in.
Diameter (English) Interior 0.37 in.
Diameter (Metric) Interior 0.95 cm