Learn More
Supertek™ Polypropylene PCR Tube
GREENER_CHOICE
Catalog No. S09606
Change view
Click to view available options
Volume (Metric):
0.2 mL
Catalog No. Volume (Metric)
S09606 0.2 mL
1 options
Catalog No. S09606 Supplier Supertek™ Supplier No. A10279/1
Only null left
Thin uniform wall tube for efficient thermal transfer.
  • PCR tube made of prime virgin polypropylene
  • Transparent, soft and robust
  • High light transmittance for optimal performance
Specifications
Color Transparent
Volume (Metric) 0.2 mL
Shape Cylindrical
Sterility Non-sterile
Material Polypropylene
Closure Type Optical (Attached)
Autoclavable Autoclavable
Closure Included Yes
Disposable Limited-use
Graduated No
Bottom Shape Conical
Product Type PCR Tube
Show More Show Less