missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Superior Touch™ Dyneema™ Gloves, Polyurethane Coating

Catalog No. 17400171
Change view
Click to view available options
Size:
11
5
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17400171 5
17400172 6
17400173 7
296361989 8
296360724 9
296361985 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17400171 Supplier Superior Glove™ Supplier No. S13SXGPUQ/5
Only null left

Cut-resistant gloves with Dyneema™ filament yarn feature a polyurethane coated palm for tacky grip in dry conditions.

  • Polyurethane palm coating is excellent for a 'tacky' grip in dry conditions
  • Lint-free continuous filament yarn
  • High dexterity and flexibility
  • Gloves are washable and can be bleached, at cold temperatures

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 5
For Use With (Application) Sharp steel, HVAC, glass metal fabrication, circuit boards, automotive
Glove Cuff Style Knit
Model S13SXGPUQ/5
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Yarn Gauge 13
Show More Show Less