missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Superior Touch™ Dyneema™ Gloves, Foam Nitrile Palm

Catalog No. 296359021
Change view
Click to view available options
Size:
7
8
9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296359021 7
296359022 8
296359023 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296359021 Supplier Superior Glove™ Supplier No. S13SXGBFN7
Only null left

Speckled gray Dyneema™ cut-resistant gloves feature a foam nitrile palm for secure grip when working with liquids and oils.

  • Speckled gray, 13-gauge gloves
  • Made of knit with lint-free, continuous-filament Dyneema™
  • Foam nitrile improves grip by absorbing and displacing liquids and oils on smooth objects
  • Gloves are washable and can be bleached at cold temperatures only

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) sharp Steel, Glass, circuit boards, automotive
Glove Cuff Style Knit
Model S13SXGBFN7
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Superior Touch™
Yarn Gauge 13
Show More Show Less