missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Superior Glove™ Superior Touch™ Cut-Resistant Composite-Knit Gloves

$189.30 - $189.30

Specifications

Color Gray
Cut Resistance Rating Level 4
Material Dyneema™,Silica Infused Fiber,Spandex™
For Use With (Application) meat processing, food and beverage, handling sharp materials, solar panel manufacturing
Glove Cuff Style Knit
View More Specs

Products 3
Catalog Number Mfr. No. Size Hem Color Price Quantity  
Catalog Number Mfr. No. Size Hem Color Price Quantity  
17401266 View Documents Superior Glove™
S13GDSTLS
Small Red Pack of 12 Pair for $189.30
Case of 20 Pack for $3,621.40
 
17401267 View Documents Superior Glove™
S13GDSTLM
Medium Yellow Pack of 12 Pair for $189.30
Case of 20 Pack for $3,621.40
 
17401268 View Documents Superior Glove™
S13GDSTLL
Large White Pack of 12 Pair for $189.30
Case of 20 Pack for $3,621.40
 
Specifications

Specifications

Gray
Dyneema™,Silica Infused Fiber,Spandex™
Knit
Level 4
meat processing, food and beverage, handling sharp materials, solar panel manufacturing