missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Superior Touch™ Polyurethane Coated Seamless Knit Dyneema™ Fiber Gloves

Catalog No. 29635472
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29635472 8
296351453 6
29635471 7
29635473 9
29635474 10
296352572 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 29635472 Supplier Superior Glove™ Supplier No. S13SXPU8
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dyneema™ construction provides lint-free cut and abrasion protection. Polyurethane coating on the palm enhances dexterity and tactile sensitivity, while also enhancing comfort and grip.

  • Dyneema™ construction provides lint-free cut and abrasion protection
  • Polyurethane coating on the palm enhances dexterity and tactile sensitivity
  • Increased grip capabilities in dry, or slightly oily conditions
  • CFIA approved for food handling

Specifications

Coated Area Palm
Construction Seamless Knit
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A2
Hand Specific
Material Dyneema
Product Type Seamless Knit Gloves
Size 8
For Use With (Application) Circuit Boards, Glass Handling, Sharp Steel, Food Handling, and Metal Fabrication
Glove Cuff Style Knit
Series S13SXPU
Yarn Gauge 13
Show More Show Less