missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Chemstop™ Fully Nitrile Coated Cotton Gloves

Catalog No. 296355207
Change view
Click to view available options
Size:
10
7
9
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296355207 10
296355204 7
296355206 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296355207 Supplier Superior Glove™ Supplier No. GNTFC10
Edge

Nitrile rubber coating is resistant to oils and grease for enhanced grip capabilities.

    Nitrile rubber coating covers the entire glove surface
  • Gloves are resistant to grease and oil for enhanced grip capabilities
  • Knit wrist prevents dirt and debris from entering the glove

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue, White
Hand Specific
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Brick Laying, Castings, Metal Stamping, Sheet Metal, and Plywood
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series GNTFC