missing translation for 'onlineSavingsMsg'
Learn More

Superior Glove™ Chemstop™ Nitrile Coated Cotton Gloves

Catalog No. 296358700
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296358700 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296358700 Supplier Superior Glove™ Supplier No. GNT10
Only null left

Nitrile rubber coating is resistant to oils and grease for enhanced grip capabilities.

    Nitrile rubber coating covers 3/4 of the glove surface
  • Gloves are resistant to grease and oil for enhanced grip capabilities
  • Knit wrist prevents dirt and debris from entering the glove

Specifications

ANSI Abrasion Rating ANSI 2
Coating Material Nitrile
Color Blue
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Brick Laying, Castings, and Metal Stamping
Glove Cuff Color Natural
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series Chemstop™ GNT
Show More Show Less